3,4-Dichloroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
437778-250G 437778-5G
SIAL437778-250GEA 82.9 EUR
SIAL437778-250G SIAL437778-5G
3,4-Dichloroaniline, Sigma-Aldrich®
3,4-Dichloroaniline
Formula: C₆H₅Cl₂N
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 70…73 °C
Density: 1.567 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007768
CAS Number: 95-76-1
EINECS: 202-448-4
UN: 3442
ADR: 6.1,II
Merck Index: 13,03079