4-Chloroaniline, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
477222-1G
477222-10G
SIAL477222-1GEA
42.4
EUR
SIAL477222-1G
SIAL477222-10G
4-Chloroaniline, Sigma-Aldrich®
4-Chloroaniline
Formula:
C₆H₆ClN MW: 127.57 g/mol Boiling Pt: 232 °C (1013 hPa) Melting Pt: 69…71 °C Density: 1.43 g/cm³ (20 °C) Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007835 CAS Number: 106-47-8 EINECS: 203-401-0 UN: 2018 ADR: 6.1,II Merck Index: 13,02136 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...