4-Chloroaniline, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
C22415-100G C22415-1KG C22415-500G C22415-5G
SIALC22415-100GEA 26.5 EUR
SIALC22415-100G SIALC22415-1KG SIALC22415-500G SIALC22415-5G
4-Chloroaniline, Sigma-Aldrich®
4-Chloroaniline
Formula: C₆H₆ClN
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 69…71 °C
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 13,02136