2,3,4-Trichloronitrobenzene
Supplier: Thermo Scientific
Synonyms:
1-Nitro-2,3,4-trichlorobenzene, 1,2,3-Trichloro-4-nitrobenzene, 4-Nitro-1,2,3-trichlorobenzene
SB01382.1
SB01382EA
SB01382.50
MAYBSB01382.1EA
0
EUR
MAYBSB01382.1
MAYBSB01382EA
MAYBSB01382.50
2,3,4-Trichloronitrobenzene
2,3,4-Trichloronitrobenzene
Formula:
C₆H₂Cl₃NO₂ MW: 226.45 g/mol Melting Pt: 55.5 °C |
MDL Number:
MFCD00007063 CAS Number: 17700-09-3 EINECS: 241-705-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...