4-Chloro-3-nitrophenol, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

361127-25G 361127-5G
SIAL361127-25GEA 160 EUR
SIAL361127-25G SIAL361127-5G
4-Chloro-3-nitrophenol, Sigma-Aldrich®
4-Chloro-3-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 125…128 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00043546
CAS Number: 610-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR