1-Chloro-2,4-dinitrobenzene, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
138630-500G 138630-5G
SIAL138630-500GEA 127 EUR
SIAL138630-500G SIAL138630-5G
1-Chloro-2,4-dinitrobenzene, Sigma-Aldrich®
1-Chloro-2,4-dinitrobenzene
Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 52…54 °C
Density: 1.7 g/cm³ (20 °C)
Flash Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007075
CAS Number: 97-00-7
EINECS: 202-551-4
UN: 3441
ADR: 6.1,II
Merck Index: 13,02155