1-Chloro-2,4-dinitrobenzene, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

237329-10G 237329-50G
SIAL237329-10GEA 41.4 EUR
SIAL237329-10G SIAL237329-50G
1-Chloro-2,4-dinitrobenzene, Sigma-Aldrich®
1-Chloro-2,4-dinitrobenzene
Formula: C₆H₃ClN₂O₄
MW: 202.55 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 52…54 °C
Density: 1.7 g/cm³ (20 °C)
Flash Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007075
CAS Number: 97-00-7
EINECS: 202-551-4
UN: 3441
ADR: 6.1,II
Merck Index: 13,02155

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR