1-Chloro-2,5-dimethoxybenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23497.18
B23497.30
B23497.18EA
58.4
EUR
B23497.18
B23497.30
1-Chloro-2,5-dimethoxybenzene ≥99%
1-Chloro-2,5-dimethoxybenzene
Purity: 99%
CAS No.: 2100-42-7
Molecular Formula: C8H9ClO2
Formula Weight: 172.61
Flash Point:
Formula:
C₈H₉ClO₂ Boiling Pt: 231…234 °C Melting Pt: 8.1 °C Density: 1.210 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008356 CAS Number: 2100-42-7 EINECS: 218-267-9 |