2-Chlorophenol ≥98%, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Chloro-2-hydroxybenzene
M-Clarity™ quality level = MQ100
C62794-100G C62794-1KG C62794-2KG
SIALC62794-100GEA 59.2 EUR
SIALC62794-100G SIALC62794-1KG SIALC62794-2KG
2-Chlorophenol ≥98%, Sigma-Aldrich®
2-Chlorophenol
Formula: C₆H₅ClO
MW: 128.56 g/mol
Boiling Pt: 175
Melting Pt: 9 °C
Density: 1.25 g/cm³ (20 °C)
Flash Pt: 72
Storage Temperature: Ambient
MDL Number: MFCD00002159
CAS Number: 95-57-8
EINECS: 202-433-2
UN: 2021
ADR: 6.1 III
Merck Index: 12,02173