3-Chlorophenol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
C62808-100G C62808-25G
SIALC62808-100GEA 69.6 EUR
SIALC62808-100G SIALC62808-25G
3-Chlorophenol, Sigma-Aldrich®
3-Chlorophenol
Formula: C₆H₅ClO
MW: 128.56 g/mol
Boiling Pt: ∼214 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1.245 g/cm³ (20 °C)
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002256
CAS Number: 108-43-0
EINECS: 203-582-6
UN: 2020
ADR: 6.1,III
Merck Index: 13,02173