4-Chlorophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: p-Chlorophenol
M-Clarity™ quality level = MQ200
185787-100G 185787-500G 185787-5G
SIAL185787-100GEA 38 EUR
SIAL185787-100G SIAL185787-500G SIAL185787-5G
4-Chlorophenol, Sigma-Aldrich®
4-Chlorophenol
Formula: C₆H₅ClO
MW: 128.56 g/mol
Boiling Pt: 220
Melting Pt: 41…44 °C
Density: 1.31 g/cm³ (20 °C)
Flash Pt: 121
Storage Temperature: Ambient
MDL Number: MFCD00002318
CAS Number: 106-48-9
EINECS: 203-402-6
UN: 2020
ADR: 6.1 III
Merck Index: 13,02173