2,4,5-Trichlorophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL156515-100GEA 217 EUR
SIAL156515-100G SIAL156515-25G
2,4,5-Trichlorophenol, Sigma-Aldrich®
2,4,5-Trichlorophenol
Formula: C₆H₃Cl₃O
MW: 197,45 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 68 °C
Density: 1,68 g/cm³ (20 °C)
MDL Number: MFCD00002170
UN: 3077
ADR: 9,III
Merck Index: 12,09772