2,4-Dichlorothiophenol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2,4-Dichlorobenzenethiol, 1,3-Dichloro-4-mercaptobenzene, 2,4-Dichlorobenzene-1-thiol

M-Clarity™ quality level = MQ200

540862-1G 540862-5G
SIAL540862-1GEA 31.8 EUR
SIAL540862-1G SIAL540862-5G
2,4-Dichlorothiophenol, Sigma-Aldrich®
2,4-Dichlorothiophenol
Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 115 °C (8 mmHg)
Melting Pt: 22 °C
Density: 1.44 g/cm³ (20 °C)
MDL Number: MFCD00041421
CAS Number: 1122-41-4
EINECS: 214-346-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR