2,3-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,3-Dichlorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ200

546917-25G 546917-5G
SIAL546917-25GEA 94.2 EUR
SIAL546917-25G SIAL546917-5G
2,3-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
2,3-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Boiling Pt: 140…142 °C (2 mmHg)
Melting Pt: 61…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051844
CAS Number: 82417-45-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR