2,5-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,5-Dichlorobenzenesulphonyl Chloride, 2,5-Dichlorobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
177547-100G 177547-25G
SIAL177547-100GEA 413 EUR
SIAL177547-100G SIAL177547-25G
2,5-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
2,5-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Melting Pt: 39 °C
MDL Number: MFCD00007429
CAS Number: 5402-73-3
EINECS: 226-453-6
UN: 3261
ADR: 8,II