2-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Chlorobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
546925-25G 546925-5G
SIAL546925-25GEA 169 EUR
SIAL546925-25G SIAL546925-5G
2-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®
2-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
MW: 211.07 g/mol
Boiling Pt: 88…92 °C (0.1 mmHg)
Melting Pt: 28 °C
Density: 1.548 g/cm³ (20 °C)
MDL Number: MFCD00051520
CAS Number: 2905-23-9
EINECS: 220-807-3
UN: 3265
ADR: 8,II