3-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Chlorobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ200
546968-25G 546968-5G
SIAL546968-25GEA 144 EUR
SIAL546968-25G SIAL546968-5G
3-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®
3-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
MW: 211.07 g/mol
MDL Number: MFCD00051697
CAS Number: 2888-06-4
EINECS: 220-753-0
UN: 3265
ADR: 8 II