4-Chlorobenzenesulphonyl chloride

Supplier: Thermo Scientific

Synonyms: 4-Chlorobenzene-1-sulphonyl chloride

SB00912.1 SB00912.10 SB00912.50
MAYBSB00912.1EA 0 EUR
MAYBSB00912.1 MAYBSB00912.10 MAYBSB00912.50
4-Chlorobenzenesulphonyl chloride
4-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
MW: 211.07 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00007439
CAS Number: 98-60-2
EINECS: 202-685-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR