2-Cyano-5-methoxybenzoic acid
Supplier: Thermo Scientific
BTB02971.1
BTB02971.10
MAYBBTB02971.1EA
0
EUR
MAYBBTB02971.1
MAYBBTB02971.10
2-Cyano-5-methoxybenzoic acid
2-Cyano-5-methoxybenzoic acid
Formula:
C₉H₇NO₃ MW: 177.16 g/mol |
MDL Number:
MFCD00116264 CAS Number: 179028-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...