4-Hydroxybenzonitrile, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: p-Cyanophenol, 4-Cyanophenol, p-Hydroxybenzonitrile
M-Clarity™ quality level = MQ200
C94009-100G C94009-25G C94009-5G
SIALC94009-100GEA 66.8 EUR
SIALC94009-100G SIALC94009-25G SIALC94009-5G
4-Hydroxybenzonitrile, Sigma-Aldrich®
4-Hydroxybenzonitrile
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 148 °C (1 mmHg)
Melting Pt: 109…112 °C
Density: ∼1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002312
CAS Number: 767-00-0
EINECS: 212-175-2