3-Cyanobenzenesulphonyl chloride ≥97%
Supplier: Thermo Scientific
Danger
Synonyms: 3-Cyanobenzene-1-sulphonyl chloride, 3-Cyanobenzene-1-sulphonylchloride
L20169.03 L20169.06
L20169.03EA 91.5 EUR
L20169.03 L20169.06
3-Cyanobenzenesulphonyl chloride ≥97%
3-Cyanobenzenesulphonyl chloride
Purity: 97% CAS No.: 56542-67-7 Molecular Formula: C7H4ClNO2S Formula Weight: 201.63 Flash Point: 110°(230°F) Melting Point: 47-52° Boiling Point: 114-116°/0.2mm Beilstein Reference: 2832925 UN No.: 3261 MDL No.: MFCD00203480
Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
Boiling Pt: 114…116 °C (0.2 mmHg)
Melting Pt: 47…52 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00203480
CAS Number: 56542-67-7
UN: 3261
ADR: 8,II