3-Cyanobenzenesulphonyl chloride
Supplier: Thermo Scientific
Synonyms: 3-Cyanobenzene-1-sulphonyl chloride, 3-Cyanobenzene-1-sulphonylchloride
TL00666.1 TL00666EA
MAYBTL00666.1EA 0 EUR
MAYBTL00666.1 MAYBTL00666EA
3-Cyanobenzenesulphonyl chloride
3-Cyanobenzenesulphonyl chloride
Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
MDL Number: MFCD00203480
CAS Number: 56542-67-7
UN: 3261
ADR: 8,II