Phthalonitrile, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,2-Benzenedicarbonitrile, Benzene-1,2-dicarbonitrile, 1,2-Dicyanobenzene
M-Clarity™ quality level = MQ100
171719-100G 171719-500G
SIAL171719-100GEA 46 EUR
SIAL171719-100G SIAL171719-500G
Phthalonitrile, Sigma-Aldrich®
Phthalonitrile
Formula: C₈H₄N₂
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 138…141 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001771
CAS Number: 91-15-6
EINECS: 202-044-8
UN: 3439
ADR: 6.1,II