3,6-Dihydroxyphthalonitrile 98%
Supplier: Thermo Fisher Scientific
Danger
232050050 232050250
ACRO232050050EA 35.9 EUR
ACRO232050050 ACRO232050250
3,6-Dihydroxyphthalonitrile 98%
3,6-Dihydroxyphthalonitrile
Formula: C₈H₄N₂O₂
Melting Pt: >230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001790
CAS Number: 4733-50-0
EINECS: 225-241-0
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Brown to khaki Crystalline powder or needles
Infrared spectrum Conforms
HPLC ≥97.5 %