3,6-Dihydroxyphthalonitrile, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,3-Dicyanohydroquinone, 3,6-Dihydroxyphthalonitrile
M-Clarity™ quality level = MQ200
141135-25G 141135-5G
SIAL141135-25GEA 133 EUR
SIAL141135-25G SIAL141135-5G
3,6-Dihydroxyphthalonitrile, Sigma-Aldrich®
3,6-Dihydroxyphthalonitrile
Formula: C₈H₄N₂O₂
MW: 160.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001790
CAS Number: 4733-50-0
EINECS: 225-241-0
UN: 3439
ADR: 6.1,II