Benzoyl bromide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
139726-250G 139726-50G
SIAL139726-250GEA 748 EUR
SIAL139726-250G SIAL139726-50G
Benzoyl bromide, Sigma-Aldrich®
Benzoyl bromide
Formula: C₇H₅BrO
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1.527…1.5564 g/cm³ (10 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000112
CAS Number: 618-32-6
EINECS: 210-544-2
UN: 3265
ADR: 8,II