Benzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Benzoic acid chloride
M-Clarity™ quality level = MQ200
259950-1L 259950-250ML 259950-5ML
SIAL259950-1LEA 219 EUR
SIAL259950-1L SIAL259950-250ML SIAL259950-5ML
Benzoyl chloride, Sigma-Aldrich®
Benzoyl chloride
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 197.2 °C (1013 hPa)
Melting Pt: ∼–0.6 °C
Density: 1.2023 g/cm³ (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00000653
CAS Number: 98-88-4
EINECS: 202-710-8
UN: 1736
ADR: 8,II
Merck Index: 13,01113