4-(Dimethylamino)benzoyl chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
526118-1G 526118-5G
SIAL526118-1GEA 51.3 EUR
SIAL526118-1G SIAL526118-5G
4-(Dimethylamino)benzoyl chloride, Sigma-Aldrich®
4-(Dimethylamino)benzoyl chloride
Formula: C₉H₁₀ClNO
Boiling Pt: 180 °C (15 mmHg)
Melting Pt: 147…148 °C
MDL Number: MFCD00236039
CAS Number: 4755-50-4
UN: 3261
ADR: 8,II