4-Nitrobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: p-Nitrobenzoyl chloride
M-Clarity™ quality level = MQ200
112208-250G 112208-500G 112208-50G
SIAL112208-250GEA 82.3 EUR
SIAL112208-250G SIAL112208-500G SIAL112208-50G
4-Nitrobenzoyl chloride, Sigma-Aldrich®
4-Nitrobenzoyl chloride
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 278
Melting Pt: 97…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00007345
CAS Number: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: 8 II
Merck Index: 12,06687