3,5-Dinitrobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
156272-25G 156272-100G
SIAL156272-25GEA 62.5 EUR
SIAL156272-25G SIAL156272-100G
3,5-Dinitrobenzoyl chloride, Sigma-Aldrich®
3,5-Dinitrobenzoyl chloride
Formula: C₇H₃ClN₂O₅
MW: 230.56 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007248
CAS Number: 99-33-2
EINECS: 202-750-6
UN: 3261
ADR: 8,II
Merck Index: 13,03305