Terephthaloyl dichloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Terephthaloyl chloride
M-Clarity™ quality level = MQ200
120871-1KG 120871-250G 120871-5G
SIAL120871-1KGEA 114 EUR
SIAL120871-1KG SIAL120871-250G SIAL120871-5G
Terephthaloyl dichloride, Sigma-Aldrich®
Terephthaloyl dichloride
Formula: C₈H₄Cl₂O₂
MW: 203.02 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 82…83 °C
Density: 1.34 g/cm³ (100 °C)
Flash Pt: 180 °C
MDL Number: MFCD00000693
CAS Number: 100-20-9
EINECS: 202-829-5
UN: 3261
ADR: 8,II