Salicylamide ≥98%
Supplier: Thermo Fisher Scientific
Warning
A10797.0B A10797.30
A10797.0BEA 81.4 EUR
A10797.0B A10797.30
Salicylamide ≥98%
Salicylamide
Purity: 98% CAS No.: 65-45-2 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: 138-142° Boiling Point: 181°/14mm Merck Reference: 14,8328 MDL No.: MFCD00007978
Formula: C₇H₇NO₂
Boiling Pt: 181 °C (14 mmHg)
Melting Pt: 138…142 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007978
CAS Number: 65-45-2
EINECS: 200-609-3
Merck Index: 14,8328