Anthranilohydrazide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Aminobenzoic hydrazide, 2-Aminobenzoic acid hydrazide, 2-Aminobenzohydrazide, 2-Aminobenzoylhydrazide, Anthraniloyl Hydrazine, 2-Aminobenzene-1-carbohydrazide, 2-Aminobenzhydrazide

M-Clarity™ quality level = MQ100

565490-10G 565490-50G
SIAL565490-10GEA 64.3 EUR
SIAL565490-10G SIAL565490-50G
Anthranilohydrazide, Sigma-Aldrich®
Anthranilohydrazide
Formula: C₇H₉N₃O
MW: 151.17 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 123 °C
MDL Number: MFCD00017056
CAS Number: 1904-58-1
EINECS: 217-604-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR