p-Anisohydrazide

Supplier: Thermo Scientific

Synonyms: 4-Methoxybenzoic acid hydrazide, 4-Methoxybenzoic hydrazide, 4-Methoxybenzhydrazide, 4-Methoxybenzohydrazide, 4-Methoxybenzene-1-carbohydrazide, 4-Anisohydrazide

BTB00865.1 BTB00865.10 BTB00865.50
MAYBBTB00865.1EA 0 EUR
MAYBBTB00865.1 MAYBBTB00865.10 MAYBBTB00865.50
p-Anisohydrazide
p-Anisohydrazide
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00017073
CAS Number: 3290-99-1
EINECS: 221-952-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR