4-Hydroxybenzhydrazide 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide
121000250 121001000 121005000
ACRO121000250EA 65.1 EUR
ACRO121000250 ACRO121001000 ACRO121005000
4-Hydroxybenzhydrazide 98%
4-Hydroxybenzhydrazide
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 370
Melting Pt: 260…264 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007605
CAS Number: 5351-23-5
EINECS: 226-326-5
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
HPLC ≥96.0 %