Diallyl phthalate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Phthalic acid diallyl ester

M-Clarity™ quality level = MQ100

269379-1L 269379-250ML
SIAL269379-1LEA 83.1 EUR
SIAL269379-1L SIAL269379-250ML
Diallyl phthalate, Sigma-Aldrich®
Diallyl phthalate
Formula: C₁₄H₁₄O₄
MW: 246.26 g/mol
Boiling Pt: 290
Melting Pt: –70 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 165
Storage Temperature: Ambient
MDL Number: MFCD00008646
CAS Number: 131-17-9
EINECS: 205-016-3
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR