Ethyl benzoate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Benzoic acid ethyl ester

M-Clarity™ quality level = MQ100

W242217-100G-K W242217-1KG-K W242217-25G-K
SIALW242217-100G-KEA 83.8 EUR
SIALW242217-100G-K SIALW242217-1KG-K SIALW242217-25G-K
Ethyl benzoate, Sigma-Aldrich®
Ethyl benzoate
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 415
Melting Pt: –35 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00009109
CAS Number: 93-89-0
EINECS: 202-284-3
Merck Index: 13,03802

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR