Ethyl-3-aminobenzoate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ethyl 3-aminobenzoate
M-Clarity™ quality level = MQ100
E10505-5G E10505-25G
SIALE10505-5GEA 36.9 EUR
SIALE10505-5G SIALE10505-25G
Ethyl-3-aminobenzoate, Sigma-Aldrich®
Ethyl 3-aminobenzoate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 172 °C (1013 hPa)
Density: 1.173 g/cm³ (20 °C)
MDL Number: MFCD00007794
CAS Number: 582-33-2
EINECS: 209-482-9