Benzocaine (ethyl-4-aminobenzoate), white crystalline powder
Supplier: MP Biomedicals
Warning
Synonyms: Ethyl 4-aminobenzoate, Aethoform
0215796790 0215796780
ICNA0215796790EA 569 EUR
ICNA0215796790 ICNA0215796780
Benzocaine (ethyl-4-aminobenzoate), white crystalline powder
Benzocaine (Ethyl 4-aminobenzoate)
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 88…93 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5
Merck Index: 13,01087