Benzocaine (ethyl-4-aminobenzoate), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ethyl 4-aminobenzoate, Aethoform
M-Clarity™ quality level = MQ200
112909-100G 112909-500G 112909-5G
SIAL112909-100GEA 36.9 EUR
SIAL112909-100G SIAL112909-500G SIAL112909-5G
Benzocaine (ethyl-4-aminobenzoate), Sigma-Aldrich®
Benzocaine (Ethyl 4-aminobenzoate)
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 89…90 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007892
CAS Number: 94-09-7
EINECS: 202-303-5
Merck Index: 13,01087