Methyl-4-aminobenzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl 4-aminobenzoate
MQ100
274186-100G 274186-25G 274186-5G
SIAL274186-100GEA 160 EUR
SIAL274186-100G SIAL274186-25G SIAL274186-5G
Methyl-4-aminobenzoate, Sigma-Aldrich®
Methyl 4-aminobenzoate
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 299
Melting Pt: 106…107 °C
Density: 1.216 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007891
CAS Number: 619-45-4
EINECS: 210-598-7