Ethyl salicylate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ethyl 2-hydroxybenzoate
M-Clarity™ quality level = MQ300
W245801-10KG-K W245801-1KG-K W245801-25KG-K
SIALW245801-10KG-KEA 351 EUR
SIALW245801-10KG-K SIALW245801-1KG-K SIALW245801-25KG-K
Ethyl salicylate, Sigma-Aldrich®
Ethyl salicylate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002215
CAS Number: 118-61-6
EINECS: 204-265-5
Merck Index: 13,03881