Methyl salicylate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

M-Clarity™ quality level = MQ100

M6752-250ML M6752-1L M6752-500ML M6752-1GA
SIALM6752-250MLEA 43.6 EUR
SIALM6752-250ML SIALM6752-1L SIALM6752-500ML SIALM6752-1GA
Methyl salicylate, Sigma-Aldrich®
Methyl salicylate
Formula: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 432
Melting Pt: 100…101 °C
Density: 1.184 g/cm³ (20 °C)
Flash Pt: 99
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR