Methyl salicylate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W274518-10KG-K W274518-1KG-K W274518-5KG-K
SIALW274518-10KG-KEA 186 EUR
SIALW274518-10KG-K SIALW274518-1KG-K SIALW274518-5KG-K
Methyl salicylate, Sigma-Aldrich®
Methyl salicylate
Formula: 2-(HO)C₆H₄CO₂CH₃
Boiling Pt: 432 °C (1013 hPa)
Melting Pt: 100…101 °C
Density: 1.184 g/cm³ (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM