Ethyl-3-hydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Hydroxybenzoic acid ethyl ester, Ethyl 3-hydroxybenzoate
M-Clarity™ quality level = MQ200
251208-100G 251208-25G
SIAL251208-100GEA 72.8 EUR
SIAL251208-100G SIAL251208-25G
Ethyl-3-hydroxybenzoate, Sigma-Aldrich®
Ethyl 3-hydroxybenzoate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 282
Melting Pt: 72…73 °C
Density: 1.068 g/cm³ (131 °C)
MDL Number: MFCD00002296
CAS Number: 7781-98-8
EINECS: 231-951-1