Butyl-4-hydroxybenzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben
M-Clarity™ quality level = MQ300
W220302-1KG W220302-5KG W220302-SAMPLE
SIALW220302-1KGEA 104 EUR
SIALW220302-1KG SIALW220302-5KG SIALW220302-SAMPLE
Butyl-4-hydroxybenzoate, Sigma-Aldrich®
Butyl 4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156…157 °C (3.5 mmHg)
Melting Pt: 73 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 13,01583