Dibutyl phthalate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: DBP, Phthalic acid dibutyl ester
M-Clarity™ quality level = MQ200
524980-1L 524980-25ML 524980-4L 524980-100ML
SIAL524980-1LEA 43.1 EUR
SIAL524980-1L SIAL524980-25ML SIAL524980-4L SIAL524980-100ML
Dibutyl phthalate, Sigma-Aldrich®
Dibutyl phthalate
Formula: C₁₆H₂₂O₄
MW: 278.35 g/mol
Boiling Pt: 340
Melting Pt: –35 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 171 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009441
CAS Number: 84-74-2
EINECS: 201-557-4
UN: 3082
ADR: 9 III
REACH: 01-2119493042-44
Merck Index: 13,01586
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd0889645c3647a1b051c82c9c74ea035c4e295&vid=561e5c60f906bd1eb64b32466d8325825293fda4&prid=web&clid=SAPDAM