Dimethyl phthalate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Phthalic acid dimethyl ester
M-Clarity™ quality level = MQ300
W508500-10KG W508500-1KG W508500-25KG W508500-SAMPLE
SIALW508500-10KGEA 269 EUR
SIALW508500-10KG SIALW508500-1KG SIALW508500-25KG SIALW508500-SAMPLE
Dimethyl phthalate, Sigma-Aldrich®
Dimethyl phthalate
Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Boiling Pt: 283.7
Melting Pt: 6
Density: 1.192 g/cm³ (20 °C)
Flash Pt: 146
Storage Temperature: Ambient
MDL Number: MFCD00008425
CAS Number: 131-11-3
EINECS: 205-011-6
Merck Index: 13,03281