Benzoic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W213128-100G-K W213128-1KG-K W213128-5KG-K
SIALW213128-100G-KEA 127 EUR
SIALW213128-100G-K SIALW213128-1KG-K SIALW213128-5KG-K
Benzoic acid, Sigma-Aldrich®
Benzoic acid
Formula: C₇H₆O₂
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 121…123 °C
Density: 1.316 g/cm³ (20 °C)
Flash Pt: 121 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002398
CAS Number: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM