Sodium benzoate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Benzoic acid sodium salt
109169-25G 109169-1KG 109169-5KG
SIAL109169-25GEA 16.9 EUR
SIAL109169-25G SIAL109169-1KG SIAL109169-5KG
Sodium benzoate, Sigma-Aldrich®
Sodium benzoate
Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: 410…430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654